Assay Details
Target Gene Details
Entrez Gene ID: | 18742 |
Gene Name: | paired-like homeodomain transcription factor 3 |
Gene Aliases: |
Ptx3, ak |
Location: |
Chr.19:46135294-46148325 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pitx3 | NM_008852.4 | 4 | 772 | NP_032878.1 |
| AB221559.1 | 3 | 640 | ||
| AF005772.1 | 4 | 768 | AAB87380.1 | |
| BC120844.1 | 3 | 661 | ||
| BC137810.1 | 3 | 661 |
More Information
Set Membership: |
|

Genomic Map