Assay Details
Target Gene Details
Entrez Gene ID: | 226089 |
Gene Name: | RAB6A GEF complex partner 1 |
Gene Aliases: |
C030046E11Rik, C130057E09Rik, Kiaa1432 |
Location: |
Chr.19:29522282-29605921 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ric1 | NM_001081319.1 | 22 | 3615 | NP_001074788.1 |
| AK048687.1 | 2 | 713 | ||
| AK136285.1 | 3 | 438 | BAE22914.1 | |
| AK173171.1 | 21 | 3197 | BAD32449.1 |

Genomic Map