Assay Details
Target Gene Details
Entrez Gene ID: | 72691 |
Gene Name: | calcium homeostasis modulator 2 |
Gene Aliases: |
2810048G17Rik, Fam26b |
Location: |
Chr.19:47132232-47138294 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Calhm2 | NM_133746.5 | 3 | 1266 | NP_598507.3 |
AK012923.1 | 3 | 1191 | BAC25382.1 | |
AK040739.1 | 3 | 1267 | BAE43310.1 | |
AK089278.1 | 3 | 1267 | BAC40825.1 | |
AK170068.1 | 3 | 1237 | BAE41544.1 | |
BC019197.1 | 3 | 1188 | AAH19197.1 | |
BC096564.1 | 3 | 417 | AAH96564.1 |
