Assay Details
Target Gene Details
Entrez Gene ID: | 381218 |
Gene Name: | RIKEN cDNA 4430402I18 gene |
Gene Aliases: |
AW122618, Spata6l |
Location: |
Chr.19:28845528-28970888 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4430402I18Rik |
Target Gene Details
Entrez Gene ID: | 74411 |
Gene Name: | phosphatidic acid phosphatase type 2 domain containing 2 |
Gene Aliases: |
4932443D16Rik |
Location: |
Chr.19:28963920-28966801 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppapdc2 | NM_028922.3 | 1 | 1920 | NP_083198.1 |
| AK016572.1 | 1 | 1921 | BAB30313.1 | |
| AK149181.1 | 1 | 1682 | ||
| BC052412.1 | 1 | 1887 | AAH52412.1 |
More Information
Set Membership: |
|

Genomic Map