Assay Details
Target Gene Details
Entrez Gene ID: | 245683 |
Gene Name: | kelch-like 34 |
Gene Aliases: |
Gm388 |
Location: |
Chr.X:157818435-157821060 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl34 | NM_001081667.2 | 1 | 329 | NP_001075136.1 |
| CA325145.1 | 1 | 583 |

Genomic Map