Assay Details
Target Gene Details
Entrez Gene ID: | 26900 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked |
Gene Aliases: |
8030469F12Rik, D1Pas1-rs1, Dby |
Location: |
Chr.Y:1260715-1286628 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx3y | NM_012008.2 | 17 | 2590 | NP_036138.1 |
AJ007376.1 | 17 | 2557 | CAA07483.1 | |
AK154382.1 | 17 | 2550 | BAE32549.1 | |
AK160604.1 | 4 | 1079 | BAE35907.1 | |
BC021453.1 | 17 | 2555 | AAH21453.1 |
