Assay Details
Target Gene Details
Entrez Gene ID: | 22290 |
Gene Name: | ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome |
Gene Aliases: |
Hydb, mKIAA4057 |
Location: |
Chr.Y:1085187-1245754 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uty | NM_009484.2 | 1 | 60 | NP_033510.2 |
| AF057367.1 | 1 | 40 | AAC67385.1 | |
| AK043154.1 | 1 | 60 | BAC31478.1 | |
| AK142344.1 | 1 | 79 | ||
| AK142512.1 | 1 | 82 | BAE25094.1 | |
| AK155922.1 | ||||
| AK162410.1 | BAE36901.1 | |||
| AK220463.1 | BAD90490.1 |
More Information
Set Membership: |
|

Genomic Map