Assay Details
Target Gene Details
Entrez Gene ID: | 56418 |
Gene Name: | YKT6 homolog (S. Cerevisiae) |
Gene Aliases: |
0610042I15Rik, 1810013M05Rik, AW105923 |
Location: |
Chr.11:5955758-5967780 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ykt6 | NM_019661.4 | 7 | 1764 | NP_062635.2 |
AK088286.1 | 1 | 823 | ||
AK147098.1 | 7 | 1724 | BAE27674.1 | |
AK150472.1 | 7 | 1721 | BAE29589.1 | |
AK159253.1 | 7 | 1711 | BAE34934.1 | |
BC006760.1 | 7 | 1690 | AAH06760.1 | |
BQ031168.1 |
