Assay Details
Target Gene Details
Entrez Gene ID: | 12192 |
Gene Name: | zinc finger protein 36, C3H type-like 1 |
Gene Aliases: |
AW742437, AW743212, Berg36, Brf1, D530020L18Rik, ERF1, TIS11b, cMG1 |
Location: |
Chr.12:80107760-80113013 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp36l1 | NM_007564.5 | 2 | 1005 | NP_031590.1 |
| AK036950.1 | 2 | 1006 | BAC29648.1 | |
| AK051035.1 | 2 | 1006 | BAC34506.1 | |
| AK077855.1 | 2 | 1003 | ||
| BC016621.1 | 2 | 990 | AAH16621.1 | |
| M58566.1 | 2 | 934 | AAA72948.1 |
More Information
Set Membership: |
|

Genomic Map