Assay Details
Target Gene Details
Entrez Gene ID: | 21408 |
Gene Name: | zinc finger protein 354A |
Gene Aliases: |
AW488485, Tcf17, Znf354a, kid1 |
Location: |
Chr.11:51059244-51072799 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp354a | NM_009329.3 | 5 | 592 | NP_033355.2 |
| AB221586.1 | 4 | 396 | ||
| AF184111.1 | 5 | 554 | AAF01033.1 | |
| AK030469.1 | 5 | 571 | ||
| BC050843.1 | 5 | 592 | AAH50843.1 | |
| BC087540.1 | 5 | 728 | AAH87540.1 | |
| L77247.1 | 4 | 456 | AAA96309.1 |

Genomic Map