Assay Details
Target Gene Details
Entrez Gene ID: | 12763 |
Gene Name: | cytidine monophospho-N-acetylneuraminic acid hydroxylase |
Gene Aliases: |
- |
Location: |
Chr.13:24326980-24477289 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmah | NM_001111110.2 | 17 | 9446 | NP_001104580.1 |
| NM_001284519.1 | 18 | 9608 | NP_001271448.1 | |
| NM_001284520.1 | 15 | 8963 | NP_001271449.1 | |
| NM_007717.5 | 16 | 9101 | NP_031743.3 | |
| AA122526.1 | 1 | 243 |
More Information
Set Membership: |
|

Genomic Map