Assay Details
Target Gene Details
Entrez Gene ID: | 17974 |
Gene Name: | non-catalytic region of tyrosine kinase adaptor protein 2 |
Gene Aliases: |
4833426I10Rik, Grb4, NCKbeta, Nck-2 |
Location: |
Chr.1:43445749-43570518 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nck2 | NM_010879.3 | 5 | 2702 | NP_035009.3 |
| AK014949.1 | 5 | 2696 | ||
| AK051665.1 | 5 | 2702 | BAC34709.1 | |
| BC011071.1 | 3 | 2267 | AAH11071.1 | |
| BC034255.1 | 5 | 2442 | AAH34255.1 | |
| BQ176012.1 | 1 | 88 |
More Information
Set Membership: |
|

Genomic Map