Assay Details
Target Gene Details
Entrez Gene ID: | 72899 |
Gene Name: | MACRO domain containing 2 |
Gene Aliases: |
1110033L15Rik, 2610107G07Rik, 2900006F19Rik |
Location: |
Chr.2:140395356-142392966 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Macrod2 | NM_001013802.3 | NP_001013824.2 | ||
| NM_028387.1 | 4 | 2396 | NP_082663.1 | |
| AK013491.1 | ||||
| AK029184.1 | 4 | 2376 | BAC26340.1 | |
| AK134694.1 | BAE22244.1 | |||
| AK160433.1 | BAE35786.1 | |||
| AK165589.1 | BAE38277.1 |

Genomic Map