Assay Details
Target Gene Details
Entrez Gene ID: | 56009 |
Gene Name: | Aly/REF export factor 2 |
Gene Aliases: |
C130042O11Rik, REF2, Refbp2 |
Location: |
Chr.1:171503478-171504750 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alyref2 | NM_019484.4 | 1 | 1221 | NP_062357.3 |
| AK048243.1 | 1 | 1222 | BAC33282.1 |
More Information
Set Membership: |
|

Genomic Map