Assay Details
Target Gene Details
Entrez Gene ID: | 76089 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 2 |
Gene Aliases: |
5830453M24Rik, CNRasGEF, Pdzgef1, RA-GEF-1, mKIAA0313, nRapGEP |
Location: |
Chr.3:79062516-79287244 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapgef2 | NM_001099624.2 | 24 | 4969 | NP_001093094.2 |
| AB093228.1 | 17 | 3852 | BAC41412.1 | |
| AK018008.1 | 1 | 86 | ||
| AK147368.1 | 24 | 4897 | ||
| BC067056.1 | 24 | 4969 | AAH67056.1 |
More Information
Set Membership: |
|

Genomic Map