Assay Details
Target Gene Details
Entrez Gene ID: | 66646 |
Gene Name: | ribulose-5-phosphate-3-epimerase |
Gene Aliases: |
2810429B02Rik, 5730518J08Rik |
Location: |
Chr.1:66700797-66719805 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpe | NM_025683.2 | 6 | 635 | NP_079959.2 |
| AK007735.1 | 6 | 641 | ||
| AK017774.1 | 6 | 636 | BAB30922.2 | |
| AK029088.1 | 6 | 634 | BAC26287.1 | |
| AK088152.1 | 6 | 636 | BAC40175.1 | |
| AK150008.1 | 6 | 673 | BAE29233.1 | |
| BC006953.1 | 6 | 629 | AAH06953.1 | |
| BC019126.1 | 6 | 678 | AAH19126.1 |

Genomic Map