Assay Details
Target Gene Details
Entrez Gene ID: | 13136 |
Gene Name: | CD55 antigen |
Gene Aliases: |
Daf, Daf-GPI, Daf1, GPI-DAF |
Location: |
Chr.1:130437196-130463012 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd55 | NM_010016.2 | 10 | 1665 | NP_034146.2 |
| AK160994.1 | BAE36139.1 |

Genomic Map