Assay Details
Target Gene Details
Entrez Gene ID: | 60532 |
Gene Name: | Wilms tumour 1-associating protein |
Gene Aliases: |
2810408K05Rik, 9430038B09Rik |
Location: |
Chr.17:12966798-12992808 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wtap | NM_001113533.1 | 8 | 1353 | NP_001107005.1 |
| AJ276707.1 | 6 | 1111 | CAC16790.1 | |
| AK156334.1 | 8 | 1249 | BAE33680.1 |

Genomic Map