Assay Details
Target Gene Details
Entrez Gene ID: | 100042807 |
Gene Name: | eukaryotic translation initiation factor 3, subunit J2 |
Gene Aliases: |
ENSMUSG00000043424, Eif3j-2, Gm9781, eIF3 p35, eIF3j-B |
Location: |
Chr.18:43475418-43477836 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif3j2 | NM_001256055.1 | 1 | 720 | NP_001242984.1 |
| BC080724.1 | 1 | 222 | AAH80724.1 | |
| BC080788.1 | 1 | 674 | AAH80788.1 | |
| BC108355.1 | 1 | 500 | AAI08356.1 |
More Information
Set Membership: |
|

Genomic Map