Assay Details
Target Gene Details
Entrez Gene ID: | 353211 |
Gene Name: | prune homolog 2 (Drosophila) |
Gene Aliases: |
6330414G02Rik, A230083H22Rik, A330102H22Rik, Bmcc1, mKIAA0367 |
Location: |
Chr.19:16955670-17223958 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prune2 | NM_181348.4 | 20 | 11381 | NP_851993.3 |
AK018172.1 | 1 | 100 | ||
AK170001.1 | 9 | 2952 | BAE41506.1 | |
AK172939.1 | 12 | 3107 | BAD32217.1 | |
BC095978.1 | 9 | 2996 | AAH95978.1 | |
FR693373.1 | 11 | 3107 | ||
GQ403051.1 | 11 | 3100 |
