Assay Details
Target Gene Details
Entrez Gene ID: | 68396 |
Gene Name: | N-acetyltransferase 8 (GCN5-related, putative) |
Gene Aliases: |
0610037O16Rik, CCNAT, Cml4 |
Location: |
Chr.6:85830387-85836636 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nat8 |

Genomic Map