Assay Details
Target Gene Details
Entrez Gene ID: | 19242 |
Gene Name: | pleiotrophin |
Gene Aliases: |
HARP, HB-GAM, HBBN, HBGF-8, HBNF, OSF, Osf-1, Osf1 |
Location: |
Chr.6:36714909-36811361 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptn | NM_008973.2 | NP_032999.1 | ||
| AK011346.1 | BAB27557.1 | |||
| AK011699.1 | BAB27787.1 | |||
| AK161964.1 | BAE36658.1 | |||
| BC002064.1 | AAH02064.1 | |||
| BC061695.1 | AAH61695.1 | |||
| D90225.1 | BAA14260.1 | |||
| EF437379.1 |
More Information
Set Membership: |
|

Genomic Map