Assay Details
Target Gene Details
Entrez Gene ID: | 434156 |
Gene Name: | EP300 interacting inhibitor of differentiation 2B |
Gene Aliases: |
3010005C08Rik |
Location: |
Chr.7:28277706-28280129 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eid2b | NM_001177427.1 | 1 | 57 | NP_001170898.1 |
AK044722.1 |
