Assay Details
Target Gene Details
Entrez Gene ID: | 320951 |
Gene Name: | phosphatidylserine decarboxylase |
Gene Aliases: |
9030221M09Rik |
Location: |
Chr.5:32736303-32785636 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pisd | NM_177298.3 | 8 | 1049 | NP_796272.2 |
| AK033483.1 | 8 | 1041 | BAC28313.1 | |
| AK034656.1 | 8 | 1051 | BAC28786.1 | |
| AK162028.1 | 8 | 1042 | BAE36689.1 | |
| AK162737.1 | 8 | 1050 | BAE37044.1 | |
| AK167554.1 | 7 | 991 | BAE39619.1 | |
| BC070408.1 | 8 | 1048 | AAH70408.1 | |
| BC094594.1 | 8 | 1040 | AAH94594.1 | |
| BC116695.1 | 8 | 1051 | ||
| BC117509.1 | 8 | 1051 |
More Information
Set Membership: |
|

Genomic Map