Assay Details
Target Gene Details
Entrez Gene ID: | 66617 |
Gene Name: | N-terminal Xaa-Pro-Lys N-methyltransferase 1 |
Gene Aliases: |
2610205E22Rik, AL033331, AL033332, Mettl11a, NTM1A |
Location: |
Chr.2:30807780-30823025 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntmt1 | NM_170592.2 | NP_733480.1 | ||
| AK009056.1 | ||||
| AK011888.1 | ||||
| AK011960.1 | ||||
| AK154783.1 | BAE32826.1 | |||
| AK179181.1 | ||||
| AK180063.1 | ||||
| AK184965.1 | ||||
| AK197657.1 | ||||
| AK218665.1 | ||||
| BC027220.1 | AAH27220.1 |
More Information
Set Membership: |
|

Genomic Map