Assay Details
Target Gene Details
Entrez Gene ID: | 67569 |
Gene Name: | MGAT4 family, member C |
Gene Aliases: |
9130411I17Rik, GntIVh |
Location: |
Chr.10:101681487-102391851 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat4c | NM_001162368.1 | NP_001155840.1 | ||
| NM_001162369.1 | NP_001155841.1 | |||
| NM_001205098.1 | NP_001192027.1 | |||
| AK032009.1 | ||||
| BB611341.1 | ||||
| CD774691.1 |
More Information
Set Membership: |
|

Genomic Map