Assay Details
Target Gene Details
Entrez Gene ID: | 353211 |
Gene Name: | prune homolog 2 (Drosophila) |
Gene Aliases: |
6330414G02Rik, A230083H22Rik, A330102H22Rik, Bmcc1, mKIAA0367 |
Location: |
Chr.19:16955670-17223958 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prune2 | NM_181348.4 | NP_851993.3 | ||
| AK038997.1 | BAC30198.1 | |||
| AK054190.1 | BAC35688.1 | |||
| AK165582.1 | BAE38273.1 | |||
| AK170001.1 | BAE41506.1 | |||
| AK172939.1 | BAD32217.1 | |||
| BC094224.1 | AAH94224.1 | |||
| BC095978.1 | AAH95978.1 | |||
| FR693373.1 | ||||
| GQ403051.1 |

Genomic Map