Assay Details
Target Gene Details
Entrez Gene ID: | 228889 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 27 |
Gene Aliases: |
C86129 |
Location: |
Chr.2:167015255-167034950 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx27 | NM_153065.3 | 21 | 2345 | NP_694705.2 |
AK134274.1 | 21 | 2346 | BAE22077.1 | |
AK138442.1 | 21 | 2369 | BAE23664.1 | |
AK146469.1 | 21 | 2373 | ||
AK205954.1 | 2 | 95 | ||
AK218996.1 | 2 | 117 | ||
BC011321.1 | 20 | 2222 | AAH11321.1 | |
BC026381.1 | 6 | 534 | AAH26381.1 |
