Assay Details
Target Gene Details
Entrez Gene ID: | 67839 |
Gene Name: | G-protein signalling modulator 1 (AGS3-like, C. elegans) |
Gene Aliases: |
1810037C22Rik, AW107933, Ags3 |
Location: |
Chr.2:26314606-26348237 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpsm1 | NM_001199146.1 | NP_001186075.1 | ||
NM_153410.5 | NP_700459.2 | |||
AK048691.1 | BAC33422.1 | |||
AK082985.1 | ||||
AK084631.1 | BAC39236.1 | |||
CA317518.1 | 1 | 25 |
