Assay Details
Target Gene Details
Entrez Gene ID: | 75801 |
Gene Name: | RIKEN cDNA 4930447C04 gene |
Gene Aliases: |
4921504I02Rik, A930035O15Rik, Six6OS, Six6as, Six6os1 |
Location: |
Chr.12:72880983-72940083 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930447C04Rik | NM_029444.2 | NP_083720.1 | ||
| AK014811.1 | BAB29563.1 | |||
| AK015397.1 | BAB29829.1 | |||
| AK020932.1 | BAB32260.1 | |||
| AY576418.1 | ||||
| BC137875.1 |

Genomic Map