Assay Details
Target Gene Details
Entrez Gene ID: | 94213 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 50 |
Gene Aliases: |
4933429B04Rik, 8430408E17Rik, GU2, RH-II |
Location: |
Chr.10:62616015-62651606 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx50 | NM_053183.2 | NP_444413.1 | ||
AF334104.1 | AAK29403.1 | |||
AK030239.1 | ||||
AK041281.1 | BAC30888.1 | |||
AK162347.1 | ||||
BC090996.1 | AAH90996.1 |
