Assay Details
Target Gene Details
Entrez Gene ID: | 11465 |
Gene Name: | actin, gamma, cytoplasmic 1 |
Gene Aliases: |
AL023024, Actg, Actl, E51 |
Location: |
Chr.11:120345689-120348491 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actg1 | NM_009609.2 | NP_033739.1 | ||
| AF195094.1 | AAF08293.1 | |||
| AK012251.1 | ||||
| AK076081.1 | BAC36167.1 | |||
| AK087983.1 | BAC40075.1 | |||
| AK152202.1 | BAE31031.1 | |||
| AK153446.1 | BAE32001.1 | |||
| AK157726.1 | BAE34171.1 | |||
| AK159867.1 | BAE35441.1 | |||
| AK162157.1 | BAE36758.1 | |||
| AK179650.1 | ||||
| AK183723.1 | ||||
| AK185831.1 | ||||
| AK217011.1 | ||||
| AK218702.1 | ||||
| BC003337.1 | AAH03337.1 | |||
| BC021796.1 | AAH21796.1 | |||
| BC023248.1 | AAH23248.1 | |||
| BC099371.1 | AAH99371.1 | |||
| M21495.1 | AAA37168.1 | |||
| X13055.1 | CAA31455.1 |
More Information
Set Membership: |
|

Genomic Map