Assay Details
Target Gene Details
Entrez Gene ID: | 18114 |
Gene Name: | ribosomal RNA processing 1 homolog (S. cerevisiae) |
Gene Aliases: |
AL033305, AL033306, AU018604, NNP-1, Nnp1 |
Location: |
Chr.10:78400362-78413043 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrp1 | NM_010925.2 | 8 | 749 | NP_035055.2 |
AF312394.1 | 7 | 512 | AAG30293.1 | |
AK077540.1 | 8 | 674 | ||
BC022910.1 | 7 | 615 | ||
BC065993.1 | 8 | 711 | AAH65993.1 | |
BC065994.1 | 8 | 669 | AAH65994.1 | |
U79773.1 | 8 | 701 | AAC53286.1 | |
U79774.1 | 8 | 701 | AAB63261.1 |
