Assay Details
Target Gene Details
Entrez Gene ID: | 140497 |
Gene Name: | cDNA sequence AF251705 |
Gene Aliases: |
CLM4, Cd300c, Cd300d, DIgR1, Igsf7, Lmir2, MAIR-II |
Location: |
Chr.11:114996721-115004357 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AF251705 | NM_134158.1 | 2 | 441 | NP_598919.1 |
| AB091767.1 | 2 | 172 | BAC77076.1 | |
| AB091768.1 | 2 | 155 | BAC77077.1 | |
| AB098476.1 | 2 | 108 | BAC80269.1 | |
| AF251705.1 | 2 | 529 | AAM19098.1 | |
| AI060397.1 | 2 | 197 | ||
| AK050008.1 | 2 | 169 | BAC34030.1 | |
| AK079395.1 | 2 | 257 | BAC37631.1 | |
| AK154808.1 | 2 | 161 | BAE32844.1 | |
| AY048685.1 | 2 | 441 | AAL12222.1 | |
| AY457050.1 | 2 | 108 | AAR27941.1 | |
| BC006801.1 | 2 | 172 | AAH06801.1 | |
| BC111820.1 | 2 | 107 | AAI11821.1 |
More Information
Set Membership: |
|

Genomic Map