Assay Details
Target Gene Details
Entrez Gene ID: | 19175 |
Gene Name: | proteasome (prosome, macropain) subunit, beta type 6 |
Gene Aliases: |
Lmp19, Mpnd |
Location: |
Chr.11:70525357-70527858 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmb6 | NM_008946.4 | 6 | 770 | NP_032972.3 |
| AK078437.1 | BAC37272.1 | |||
| AK132042.1 | 6 | 753 | BAE20959.1 | |
| AK167123.1 | BAE39271.1 | |||
| AK167227.1 | BAE39351.1 | |||
| AV072442.1 | ||||
| BC013897.1 | AAH13897.1 | |||
| BM202854.2 | 1 | 267 | ||
| U13393.1 | AAA75375.1 |
More Information
Set Membership: |
|

Genomic Map