Assay Details
Target Gene Details
Entrez Gene ID: | 12983 |
Gene Name: | colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage) |
Gene Aliases: |
AI848964, AIC2B, Bc, CDw131, Csf2rb1, Csfgmrb, Il3r, Il3rb, Il3rb1, Il5rb |
Location: |
Chr.15:78325797-78351001 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csf2rb | NM_007780.4 | 11 | 1636 | NP_031806.3 |
| AF483496.1 | 10 | 1378 | AAL90770.1 | |
| AF483497.1 | 10 | 1378 | AAL90771.1 | |
| AK152608.1 | 11 | 1576 | BAE31354.1 | |
| AK154286.1 | 11 | 1573 | ||
| AK155109.1 | 11 | 1607 | BAE33054.1 | |
| AK155215.1 | 11 | 1516 | BAE33126.1 | |
| AK155699.1 | 11 | 1573 | BAE33394.1 | |
| AK155803.1 | 11 | 1573 | BAE33441.1 | |
| AK155822.1 | 11 | 1576 | ||
| AK155885.1 | 11 | 1577 | ||
| AK171477.1 | 11 | 1573 | BAE42480.1 | |
| BC113202.1 | 12 | 1647 | AAI13203.1 | |
| M34397.1 | 12 | 1665 | AAA37204.1 | |
| S78451.1 | 4 | 266 | AAB34209.1 |
More Information
Set Membership: |
|

Genomic Map