Assay Details
Target Gene Details
Entrez Gene ID: | 14942 |
Gene Name: | granzyme E |
Gene Aliases: |
CCP3, Ctla-6, Ctla6, MCSP-2 |
Location: |
Chr.14:56117619-56120625 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gzme | NM_010373.3 | NP_034503.2 | ||
| BG146331.1 | ||||
| J03256.1 | AAA37737.1 | |||
| M36901.1 | AAA37487.1 | |||
| X14093.1 | CAA32255.1 |
More Information
Set Membership: |
|

Genomic Map