Assay Details
Target Gene Details
Entrez Gene ID: | 75329 |
Gene Name: | activating transcription factor 7 interacting protein 2 |
Gene Aliases: |
4930558K11Rik, BC018510, Get-1, PSM2 |
Location: |
Chr.16:10192640-10249669 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf7ip2 | NM_029253.1 | NP_083529.1 | ||
| NM_153123.2 | NP_694763.2 | |||
| AK016180.1 | BAB30138.1 | |||
| AK049401.1 | ||||
| AK077229.1 | BAC36698.1 | |||
| AK139862.1 | BAE24164.1 | |||
| AK145634.1 | BAE26553.1 | |||
| AY903215.1 | AAX99226.1 | |||
| BY715784.1 |
More Information
Set Membership: |
|

Genomic Map