Assay Details
Target Gene Details
Entrez Gene ID: | 13137 |
Gene Name: | decay accelerating factor 2 |
Gene Aliases: |
AI323748, Cd55b, Daf, Daf-TM, TM-DAF |
Location: |
Chr.1:130388528-130423037 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Daf2 | NM_007827.2 | NP_031853.2 | ||
| BC127150.1 | ||||
| L41365.1 | AAB00092.1 |
More Information
Set Membership: |
|

Genomic Map