Assay Details
Target Gene Details
Entrez Gene ID: | 56695 |
Gene Name: | paroxysmal nonkinesiogenic dyskinesia |
Gene Aliases: |
2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2 |
Location: |
Chr.1:74284597-74353692 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnkd | NM_001039509.1 | NP_001034598.1 | ||
NM_025580.2 | NP_079856.2 | |||
AF318058.1 | AAL25717.1 | |||
AK009545.1 | BAB26351.1 | |||
AK012976.1 | BAB28577.2 | |||
AK166548.1 | BAE38846.1 | |||
AK173125.1 | BAD32403.1 | |||
AK179109.1 | ||||
AK181664.1 | ||||
AY039041.1 | AAK83447.1 | |||
AY299972.1 | AAP58373.1 | |||
BC008274.1 | AAH08274.1 | |||
BC058945.1 | AAH58945.1 | |||
BC116236.1 | ||||
BC116237.1 | ||||
CK623847.1 |
