Assay Details
Target Gene Details
Entrez Gene ID: | 12946 |
Gene Name: | complement component (3b/4b) receptor 1-like |
Gene Aliases: |
Crry, Mcp, mCRY |
Location: |
Chr.1:195098747-195131570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cr1l | NM_013499.2 | NP_038527.2 | ||
| AI098173.1 | ||||
| AK004825.1 | BAC25098.1 | |||
| AK160440.1 | BAE35790.1 | |||
| BC028945.1 | AAH28945.1 | |||
| BC092048.1 | AAH92048.1 |

Genomic Map