Assay Details
Target Gene Details
Entrez Gene ID: | 29809 |
Gene Name: | RAB GTPase activating protein 1-like |
Gene Aliases: |
5830411O09Rik, 8430421H08Rik, 9630005B12Rik, AW049894, HHL, Hh1, mKIAA0471 |
Location: |
Chr.1:160219174-160793476 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabgap1l | NM_013862.5 | 12 | 1752 | NP_038890.3 |
| AK035796.1 | 12 | 1753 | BAC29189.1 | |
| BC145811.1 | 12 | 1597 | ||
| BC145813.1 | 12 | 1597 |
More Information
Set Membership: |
|

Genomic Map