Assay Details
Target Gene Details
Entrez Gene ID: | 22218 |
Gene Name: | SMT3 suppressor of mif two 3 homolog 1 (yeast) |
Gene Aliases: |
GMP1, PIC1, SENTRIN, SMT3, SMT3H3, SMTP3, SUMO-1, Smt3C, Ubl1 |
Location: |
Chr.1:59639434-59670834 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sumo1 | NM_009460.2 | NP_033486.1 | ||
| AF033353.1 | AAC39959.1 | |||
| AK002536.1 | BAB22172.1 | |||
| AK011074.1 | BAB27379.1 | |||
| AK089081.1 | BAC40739.1 | |||
| AK159366.1 | BAE35024.1 | |||
| AK220052.1 | ||||
| BC082566.1 | AAH82566.1 | |||
| BC083158.1 | AAH83158.1 | |||
| BY242724.1 |

Genomic Map