Assay Details
Target Gene Details
Entrez Gene ID: | 69821 |
Gene Name: | mitochondrial transcription termination factor 4 |
Gene Aliases: |
1810059A23Rik, 4933412H03Rik, Mterfd2 |
Location: |
Chr.1:93299211-93305965 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mterf4 | NM_178051.3 | NP_835152.1 | ||
AK007901.1 | ||||
AK033043.1 | ||||
AK077142.1 | BAC36640.1 | |||
AK079098.1 | ||||
BC064709.1 | AAH64709.1 | |||
BY261356.1 |
