Assay Details
Target Gene Details
Entrez Gene ID: | 16624 |
Gene Name: | kallikrein 1-related peptidase b8 |
Gene Aliases: |
Klk8, Nrpn, TADG14, mGK-8 |
Location: |
Chr.7:43950672-43954938 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klk1b8 | NM_008457.2 | NP_032483.1 | ||
| BC031798.1 | AAH31798.1 | |||
| X03994.1 | CAA27630.1 |
More Information
Set Membership: |
|

Genomic Map