Assay Details
Target Gene Details
Entrez Gene ID: | 11872 |
Gene Name: | ADP-ribosyltransferase 2b |
Gene Aliases: |
ART2.2, ARTC2, Art, Ly92b, Rt-6, Rt6, Rt6-2 |
Location: |
Chr.7:101575729-101585583 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Art2b | NM_019915.2 | NP_064299.2 | ||
| AF016463.1 | AAB71683.1 | |||
| AF016465.1 | AAB71684.1 | |||
| X87612.1 | CAA60948.1 |
More Information
Set Membership: |
|

Genomic Map