Assay Details
Target Gene Details
Entrez Gene ID: | 67861 |
Gene Name: | aldo-keto reductase family 1, member B10 (aldose reductase) |
Gene Aliases: |
2310005E10Rik, Akr1b16 |
Location: |
Chr.6:34384206-34396954 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1b10 | NM_172398.3 | NP_765986.3 | ||
| AK019906.1 | ||||
| AK075865.1 | ||||
| AK082777.1 | BAC38615.1 | |||
| AK087002.1 | ||||
| AK169490.1 | ||||
| BC026843.1 | AAH26843.1 | |||
| BC037690.1 | AAH37690.1 |
More Information
Set Membership: |
|

Genomic Map