Assay Details
Target Gene Details
Entrez Gene ID: | 69187 |
Gene Name: | endoplasmic reticulum protein 27 |
Gene Aliases: |
1810033M07Rik, 1810047B09Rik |
Location: |
Chr.6:136907314-136922461 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erp27 | NM_026983.2 | 4 | 442 | NP_081259.1 |
| AK007684.1 | 4 | 443 | BAB25188.1 | |
| AK007800.1 | 4 | 363 | BAB25265.1 | |
| AK131830.1 | 4 | 443 | BAE20822.1 | |
| BC104406.1 | 4 | 430 | AAI04407.1 | |
| BC104407.2 | 4 | 430 | AAI04408.1 |
More Information
Set Membership: |
|

Genomic Map