Assay Details
Target Gene Details
Entrez Gene ID: | 73288 |
Gene Name: | coiled-coil domain containing 132 |
Gene Aliases: |
1700034M03Rik, 8430415E05Rik, BLM, C87205, mKIAA1861 |
Location: |
Chr.6:3498393-3603531 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc132 | NM_001167750.1 | 18 | 1646 | NP_001161222.1 |
NM_001167751.1 | 18 | 1646 | NP_001161223.1 | |
NM_024260.5 | 18 | 1646 | NP_077222.4 | |
AK042031.1 | 18 | 1626 | BAC31137.1 | |
AK075898.1 | 18 | 1635 | BAC36038.1 | |
AK129458.1 | 18 | 1596 | BAC98268.1 | |
AK134533.1 | 2 | 3445 | ||
AY254697.1 | 18 | 1639 | AAO84056.1 | |
BC002304.1 | 3 | 191 | AAH02304.1 | |
BC036294.1 | 18 | 1586 | AAH36294.2 | |
BC043092.1 | 18 | 1603 | AAH43092.1 |
