Assay Details
Target Gene Details
Entrez Gene ID: | 192196 |
Gene Name: | LUC7-like 2 (S. cerevisiae) |
Gene Aliases: |
4930471C18Rik, AA522013, AU015269, CGI59, Cgi74 |
Location: |
Chr.6:38551339-38609471 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Luc7l2 | NM_001170848.1 | 1 | 239 | NP_001164319.1 |
| NM_001170849.1 | 1 | 239 | NP_001164320.1 | |
| NM_138680.2 | 1 | 239 | NP_619621.2 | |
| AF318301.1 | 1 | 164 | AAK01182.1 | |
| AK038543.1 | 1 | 153 | ||
| AK044839.1 | 1 | 240 | ||
| AK046307.1 | 1 | 350 | ||
| AK048581.1 | 1 | 153 | ||
| AK053863.1 | 1 | 153 | ||
| AK134191.1 | 1 | 153 | BAE22044.1 | |
| AK147889.1 | 1 | 170 | BAE28206.1 | |
| AK155264.1 | 1 | 229 | ||
| AK166155.1 | 1 | 195 | ||
| BC056354.1 | 1 | 237 | AAH56354.1 | |
| BC056383.1 | 1 | 218 | AAH56383.1 | |
| BC056970.1 | 1 | 237 | AAH56970.1 |

Genomic Map